Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Tryptophanamide Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. T23125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23125G 5 g
T23121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23125G Supplier TCI America Supplier No. T23125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5022-65-1
Molecular Formula C11H14ClN3O
Molecular Weight (g/mol) 239.70
MDL Number MFCD00054315
InChI Key WOBDANBSEWOYKN-FVGYRXGTSA-N
Synonym l-tryptophanamide hydrochloride, h-trp-nh2.hcl, s-2-amino-3-1h-indol-3-yl propanamide hydrochloride, h-trp-nh2 hcl, l-tryptophane amide hydrochloride, 2s-2-amino-3-1h-indol-3-yl propanamide hydrochloride, l---tryptophan methyl ester hydrochloride, h-trp-nh hcl, h-trp-nh2 . hcl
PubChem CID 6452180
IUPAC Name (2S)-2-amino-3-(1H-indol-3-yl)propanamide hydrochloride
SMILES Cl.N[C@@H](CC1=CNC2=CC=CC=C12)C(N)=O

Specifications

Melting Point 250°C
Color White-Yellow
Quantity 5 g
Formula Weight 239.70
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material L-Tryptophanamide Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.