Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-tryptophanamide, N-((3-methyl-1H-inden-2-yl)carbonyl)-D-alanyl-N-((1S)-2-((2R)-2-methyl-2-oxiranyl | 1416709-79-9 | 98.9% | 618.72 g·mol⁻¹ | C37H38N4O5 | 10MM 1ML
SDP

Catalog No. 5000235976
Encompass_Preferred

PR-924 is a selective tripeptide epoxyketone inhibitor of the immunoproteasome subunit LMP-7, intended for research use in biochemical and cellular assays. It is supplied in ready-to-use solutions and in solid quantities to support formulation and dosing studies.

  • Selective tripeptide epoxyketone LMP-7 inhibitor.
  • Available as 10 mM solution in DMSO and multiple solid sizes.
  • High reported purity: 98.9%.
  • Molecular weight 618.72 g·mol⁻¹; formula C37H38N4O5.
  • Soluble in DMSO; in vivo formulation options described for PEG300/tween/saline and corn oil.
  • Storage recommendations provided for powder and in solvent to preserve stability.

Catalog No. 50-002-35976 Supplier Medchemexpress LLC Supplier No. HY12358710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PR-924 is a selective tripeptide epoxyketone inhibitor of the immunoproteasome subunit LMP-7, intended for research use in biochemical and cellular assays. It is supplied in ready-to-use solutions and in solid quantities to support formulation and dosing studies.

  • Selective tripeptide epoxyketone LMP-7 inhibitor.
  • Available as 10 mM solution in DMSO and multiple solid sizes.
  • High reported purity: 98.9%.
  • Molecular weight 618.72 g·mol⁻¹; formula C37H38N4O5.
  • Soluble in DMSO; in vivo formulation options described for PEG300/tween/saline and corn oil.
  • Storage recommendations provided for powder and in solvent to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.