Learn More
Medchemexpress LLC L-Tryptophanamide, N-[(3-methyl-1H-inden-2-yl)carbonyl]-D-alanyl-N-[(1S)-2-[(2R)-2-methyl-2-oxiranyl]- | 1416709-79-9 | 98.5% | 618.72 | 100 MG
SDP
Catalog No. 5000438557
Encompass_Preferred
(R)-PR-924 is an isomer of PR-924 and can be used as an experimental control. PR-924 is a selective tripeptide epoxyketone immunoproteasome subunit LMP-7 inhibitor with an IC50 of 22 nM. It covalently modifies proteasomal N-terminal threonine active sites, inhibits growth, and triggers apoptosis in multiple myeloma (MM) cells, demonstrating antitumor activities.
  • Can be used as an experimental control.
  • Selective tripeptide epoxyketone immunoproteasome subunit LMP-7 inhibitor.
  • Covalently modifies proteasomal N-terminal threonine active sites.
  • Inhibits growth and triggers apoptosis in multiple myeloma (MM) cells.
  • Exhibits antitumor activities.
Catalog No. 50-004-38557 Supplier Medchemexpress LLC Supplier No. HY123587A100MG
(R)-PR-924 is an isomer of PR-924 and can be used as an experimental control. PR-924 is a selective tripeptide epoxyketone immunoproteasome subunit LMP-7 inhibitor with an IC50 of 22 nM. It covalently modifies proteasomal N-terminal threonine active sites, inhibits growth, and triggers apoptosis in multiple myeloma (MM) cells, demonstrating antitumor activities.
  • Can be used as an experimental control.
  • Selective tripeptide epoxyketone immunoproteasome subunit LMP-7 inhibitor.
  • Covalently modifies proteasomal N-terminal threonine active sites.
  • Inhibits growth and triggers apoptosis in multiple myeloma (MM) cells.
  • Exhibits antitumor activities.