Learn More
Medchemexpress LLC L-tryptophanamide, N-((3-methyl-1H-inden-2-yl)carbonyl)-D-alanyl-N-((1S)-2-((2R)-2-methyl-2-oxiranyl)-2-oxo- | 1416709-79-9 | 98.9% | 618.72 g·mol⁻¹ | C37H38N4O5 | 5MG
SDP
Catalog No. 5000235549
Encompass_Preferred
PR-924 is a selective tripeptide epoxyketone inhibitor of the immunoproteasome β5i (LMP-7) subunit, provided as an off-white to light yellow solid for biochemical and cellular research. It shows low-nanomolar activity against LMP-7 and is supplied with a stated high purity; solubility and storage recommendations support both in vitro and in vivo use.
Catalog No. 50-002-35549 Supplier Medchemexpress LLC Supplier No. HY1235875MG
PR-924 is a selective tripeptide epoxyketone inhibitor of the immunoproteasome β5i (LMP-7) subunit, provided as an off-white to light yellow solid for biochemical and cellular research. It shows low-nanomolar activity against LMP-7 and is supplied with a stated high purity; solubility and storage recommendations support both in vitro and in vivo use.