Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Tyrosinamide | 4985-46-0 | 98.95% | 180.20 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYW008559

Encompass_Preferred

L-Tyrosinamide is a derivative of Tyrosine, which is an amino acid amide.

  • Purity of 98.95%
  • Molecular weight of 180.20
  • Appears as a solid
  • Store at 4°C under nitrogen
  • Solvent storage at -80°C for 6 months or -20°C for 1 month under nitrogen
  • Soluble in DMSO at 100 mg/mL
  • For research use only

Catalog No. 50-004-25827


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.