Learn More
Medchemexpress LLC L-Tyrosine-13C6 | 201595-63-3 | 187.14 | 5 MG
SDP
Catalog No. 5000390017
Encompass_Preferred
L-Tyrosine-13C6 is the 13C-labeled L-Tyrosine. L-Tyrosine is a non-essential amino acid which can inhibit citrate synthase activity in the posterior cortex.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • For research use only.
  • Purity: 99.20%.
Catalog No. 50-003-90017 Supplier Medchemexpress LLC Supplier No. HYN0473S25MG
L-Tyrosine-13C6 is the 13C-labeled L-Tyrosine. L-Tyrosine is a non-essential amino acid which can inhibit citrate synthase activity in the posterior cortex.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • For research use only.
  • Purity: 99.20%.