Learn More
Medchemexpress LLC L-Tyrosine-15N | 35424-81-8 | 182.18 | 25 MG
SDP
Catalog No. 5000389279
Encompass_Preferred
L-Tyrosine-15N is the 15N-labeled L-Tyrosine, a non-essential amino acid. This compound serves as a valuable tool in scientific research, particularly for quantitative analysis and as a tracer in various applications.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-89279 Supplier Medchemexpress LLC Supplier No. 25MGHYN0473S1
L-Tyrosine-15N is the 15N-labeled L-Tyrosine, a non-essential amino acid. This compound serves as a valuable tool in scientific research, particularly for quantitative analysis and as a tracer in various applications.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS