Learn More
Medchemexpress LLC L-Tyrosine-15N | 35424-81-8 | 182.18 | 5 MG
SDP
Catalog No. 5000389050
Encompass_Preferred
L-Tyrosine-15N is the 15N-labeled L-Tyrosine. L-Tyrosine is a non-essential amino acid that can inhibit citrate synthase activity in the posterior cortex.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89050 Supplier Medchemexpress LLC Supplier No. HYN0473S15MG
L-Tyrosine-15N is the 15N-labeled L-Tyrosine. L-Tyrosine is a non-essential amino acid that can inhibit citrate synthase activity in the posterior cortex.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.