Learn More
Medchemexpress LLC L-Tyrosine-17O | 104931-15-9 | 182.19 | 25 MG
SDP
Catalog No. 5000389551
Encompass_Preferred
L-Tyrosine-17O is the 17O-labeled L-Tyrosine. L-Tyrosine is a non-essential amino acid that can inhibit citrate synthase activity in the posterior cortex. It is intended for research use only and not for sale to patients. This compound is used as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89551 Supplier Medchemexpress LLC Supplier No. 25MGHYN0473S12
L-Tyrosine-17O is the 17O-labeled L-Tyrosine. L-Tyrosine is a non-essential amino acid that can inhibit citrate synthase activity in the posterior cortex. It is intended for research use only and not for sale to patients. This compound is used as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.