Learn More
Medchemexpress LLC L-Tyrosine-2,3,5,6-d4 | 62595-14-6 | 185.21 | 25 MG
SDP
Catalog No. 5000389179
Encompass_Preferred
L-Tyrosine-d4 is a deuterium-labeled L-Tyrosine. L-Tyrosine is a non-essential amino acid known to inhibit citrate synthase activity in the posterior cortex.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89179 Supplier Medchemexpress LLC Supplier No. 25MGHYN0473S
L-Tyrosine-d4 is a deuterium-labeled L-Tyrosine. L-Tyrosine is a non-essential amino acid known to inhibit citrate synthase activity in the posterior cortex.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.