Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-tyrosine-2,3,5,6-d4 | 62595-14-6 | MFCD00079605 | 98.0% | 185.21 | C9H7D4NO3 | 10 MG
SDP

Catalog No. 5000295606
Encompass_Preferred

Deuterium-labeled L-tyrosine (L-tyrosine-2,3,5,6-d4) supplied as a 10 mg solid for research use. It is intended for use as an internal standard and tracer in metabolic and isotope-labeling studies and has been reported to inhibit citrate synthase activity in the posterior cortex. Not for human or clinical use.

  • Deuterium-labeled internal standard for mass spectrometry and NMR.
  • Chemical formula C9H7D4NO3 and molecular weight 185.21 g/mol.
  • Purity (NMR) 98.0%.
  • Supplied as a 10 mg solid suitable for analytical and tracer studies.
  • For research use only; not for human or clinical applications.

Catalog No. 50-002-95606 Supplier Medchemexpress LLC Supplier No. HYN0473S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deuterium-labeled L-tyrosine (L-tyrosine-2,3,5,6-d4) supplied as a 10 mg solid for research use. It is intended for use as an internal standard and tracer in metabolic and isotope-labeling studies and has been reported to inhibit citrate synthase activity in the posterior cortex. Not for human or clinical use.

  • Deuterium-labeled internal standard for mass spectrometry and NMR.
  • Chemical formula C9H7D4NO3 and molecular weight 185.21 g/mol.
  • Purity (NMR) 98.0%.
  • Supplied as a 10 mg solid suitable for analytical and tracer studies.
  • For research use only; not for human or clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.