Learn More
Medchemexpress LLC HY-N0473 200mg Medchemexpress, L-Tyrosine CAS:60-18-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0473200MG
Encompass_Preferred
Medchemexpress, HY-N0473 200mg L-Tyrosine CAS:60-18-4 L-Tyrosine is a non-essential amino acid which can inhibit citrate synthase activity in the posterior cortex. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3588