Learn More
Medchemexpress LLC L-Tyrosine, 3-hydroxy- | 59-92-7 | 99.98% | 197.19 | 100 MG
SDP
Catalog No. 5000427699
Encompass_Preferred
L-DOPA is an orally active metabolic precursor of neurotransmitters dopamine. It can cross the blood-brain barrier and is converted into dopamine in the brain. L-DOPA has anti-allodynic effects and shows potential for Parkinson's disease research.
  • Orally active metabolic precursor of dopamine.
  • Can cross the blood-brain barrier.
  • Converted into dopamine in the brain.
  • Exhibits anti-allodynic effects.
  • Potential for Parkinson's disease treatment.
Catalog No. 50-004-27699 Supplier Medchemexpress LLC Supplier No. HYN0304R100MG
L-DOPA is an orally active metabolic precursor of neurotransmitters dopamine. It can cross the blood-brain barrier and is converted into dopamine in the brain. L-DOPA has anti-allodynic effects and shows potential for Parkinson's disease research.
  • Orally active metabolic precursor of dopamine.
  • Can cross the blood-brain barrier.
  • Converted into dopamine in the brain.
  • Exhibits anti-allodynic effects.
  • Potential for Parkinson's disease treatment.