Learn More
Medchemexpress LLC L-Tyrosine-4-13C | 81201-90-3 | 182.18 | 25 MG
SDP
Catalog No. 5000390343
Encompass_Preferred
L-Tyrosine-4-13C is a 13C-labeled L-Tyrosine. This non-essential amino acid can inhibit citrate synthase activity in the posterior cortex. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-90343 Supplier Medchemexpress LLC Supplier No. 25MGHYN0473S5
L-Tyrosine-4-13C is a 13C-labeled L-Tyrosine. This non-essential amino acid can inhibit citrate synthase activity in the posterior cortex. It is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.