Learn More
U.S. Pharmacopeia L-Tyrosine, 60-18-4, MFCD00002606, 500mg
Catalog No. NC9332031
Encompass
Molecular formula C9H11NO3, Molecular weight 181.19, Melting Point 647.6 - 651.2 °F (342 - 344 °C) (decomposes), Synonyms: Tyrosine, p-Tyrosine, 4-Hydroxy-L-phenylalanine
Catalog No. NC9332031 Supplier U.S. Pharmacopeia Supplier No. 1705006
Molecular formula C9H11NO3, Molecular weight 181.19, Melting Point 647.6 - 651.2 °F (342 - 344 °C) (decomposes), Synonyms: Tyrosine, p-Tyrosine, 4-Hydroxy-L-phenylalanine