Learn More
Medchemexpress LLC L-Tyrosine | 60-18-4 | 181.19 | 1 G
SDP
Catalog No. 5000389048
Encompass_Preferred
L-Tyrosine is a non-essential amino acid that can inhibit citrate synthase activity in the posterior cortex. It also affects other enzymes and complexes of the mitochondrial respiratory chain in various tissues like the hippocampus, striatum, and liver.
  • Inhibitors
  • Screening libraries
  • Proteins
Catalog No. 50-003-89048 Supplier Medchemexpress LLC Supplier No. HYN04731G
L-Tyrosine is a non-essential amino acid that can inhibit citrate synthase activity in the posterior cortex. It also affects other enzymes and complexes of the mitochondrial respiratory chain in various tissues like the hippocampus, striatum, and liver.
  • Inhibitors
  • Screening libraries
  • Proteins