Learn More
Medchemexpress LLC L-Tyrosine | 60-18-4 | 181.19 | 500 MG
SDP
Catalog No. 5000389904
Encompass_Preferred
L-Tyrosine is a non-essential amino acid and an endogenous metabolite. It is involved in research areas such as neurological diseases and cancer. L-Tyrosine can inhibit citrate synthase activity in the posterior cortex and has been shown to affect various enzyme activities and mitochondrial respiratory chain complexes in different tissues.
  • Non-essential amino acid
  • Endogenous metabolite
  • Involved in neurological disease research
  • Involved in cancer research
  • Inhibits citrate synthase activity
  • Affects malate dehydrogenase and succinate dehydrogenase activity
  • Modulates mitochondrial respiratory chain complexes
Catalog No. 50-003-89904 Supplier Medchemexpress LLC Supplier No. HYN0473500MG
L-Tyrosine is a non-essential amino acid and an endogenous metabolite. It is involved in research areas such as neurological diseases and cancer. L-Tyrosine can inhibit citrate synthase activity in the posterior cortex and has been shown to affect various enzyme activities and mitochondrial respiratory chain complexes in different tissues.
  • Non-essential amino acid
  • Endogenous metabolite
  • Involved in neurological disease research
  • Involved in cancer research
  • Inhibits citrate synthase activity
  • Affects malate dehydrogenase and succinate dehydrogenase activity
  • Modulates mitochondrial respiratory chain complexes