Learn More
Medchemexpress LLC L-Tyrosine-d2-1 | 30811-19-9 | 183.20 | 1 G
SDP
Supplier:  Medchemexpress LLC HYN0473S101G
Encompass_Preferred
L-Tyrosine-d2-1 is a deuterium-labeled L-Tyrosine, a non-essential amino acid. It serves as a tracer and an internal standard for quantitative analysis in techniques like NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes, such as deuterium, into drug molecules is investigated for its potential to influence pharmacokinetic and metabolic profiles during drug development. This product is stored at 4°C, protected from light and under nitrogen. In solvent, it can be stored at -80°C for 6 months or -20°C for 1 month, also protected from light and under nitrogen.
  • Used as a tracer for quantitative analysis.
  • Functions as an internal standard for analytical techniques.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Protect from light and store under nitrogen.
Catalog No. 50-003-89005