Learn More
Medchemexpress LLC L-Tyrosine-d2-1 | 30811-19-9 | 183.20 | 100 MG
SDP
Catalog No. 5000389006
Encompass_Preferred
L-Tyrosine-d2-1 is the deuterium labeled L-Tyrosine, a non-essential amino acid that can inhibit citrate synthase activity in the posterior cortex.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-89006 Supplier Medchemexpress LLC Supplier No. HYN0473S10100MG
L-Tyrosine-d2-1 is the deuterium labeled L-Tyrosine, a non-essential amino acid that can inhibit citrate synthase activity in the posterior cortex.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS