Learn More
Medchemexpress LLC L-Tyrosine-d2-1 | 30811-19-9 | 183.20 | 25 MG
SDP
Catalog No. 5000390324
Encompass_Preferred
L-Tyrosine-d2-1 is the deuterium labeled L-Tyrosine. L-Tyrosine is a non-essential amino acid which can inhibit citrate synthase activity in the posterior cortex.
  • Deuterium labeled L-Tyrosine.
  • L-Tyrosine is a non-essential amino acid.
  • Can inhibit citrate synthase activity in the posterior cortex.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-90324 Supplier Medchemexpress LLC Supplier No. 25MGHYN0473S10
L-Tyrosine-d2-1 is the deuterium labeled L-Tyrosine. L-Tyrosine is a non-essential amino acid which can inhibit citrate synthase activity in the posterior cortex.
  • Deuterium labeled L-Tyrosine.
  • L-Tyrosine is a non-essential amino acid.
  • Can inhibit citrate synthase activity in the posterior cortex.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.