Learn More
Medchemexpress LLC L-Tyrosine-d2-1 | 30811-19-9 | 183.20 | 500 MG
SDP
Catalog No. 5000389778
Encompass_Preferred
L-Tyrosine-d2-1 is the deuterium labeled L-Tyrosine. L-Tyrosine is a non-essential amino acid which can inhibit citrate synthase activity in the posterior cortex. This compound is intended for use as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development. Deuteration has gained attention for its potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled L-Tyrosine
  • Non-essential amino acid
  • Can inhibit citrate synthase activity in the posterior cortex
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • High purity: 99.30%
Catalog No. 50-003-89778 Supplier Medchemexpress LLC Supplier No. HYN0473S10500MG
L-Tyrosine-d2-1 is the deuterium labeled L-Tyrosine. L-Tyrosine is a non-essential amino acid which can inhibit citrate synthase activity in the posterior cortex. This compound is intended for use as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development. Deuteration has gained attention for its potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled L-Tyrosine
  • Non-essential amino acid
  • Can inhibit citrate synthase activity in the posterior cortex
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • High purity: 99.30%