Learn More
Medchemexpress LLC L-Tyrosine-d2-2 | 57746-15-3 | 183.20 | 1 MG
SDP
Catalog No. 5000389913
Encompass_Preferred
L-Tyrosine-d2-2 is the deuterium labeled L-Tyrosine. L-Tyrosine is a non-essential amino acid which can inhibit citrate synthase activity in the posterior cortex.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89913 Supplier Medchemexpress LLC Supplier No. HYN0473S131MG
L-Tyrosine-d2-2 is the deuterium labeled L-Tyrosine. L-Tyrosine is a non-essential amino acid which can inhibit citrate synthase activity in the posterior cortex.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.