Learn More
Medchemexpress LLC L-Tyrosine-d2 | 72963-27-0 | 183.20 | 50 MG
SDP
Catalog No. 5000389372
Encompass_Preferred
L-Tyrosine-d2 is the deuterium labeled L-Tyrosine. L-Tyrosine is a non-essential amino acid which can inhibit citrate synthase activity in the posterior cortex. This product is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89372 Supplier Medchemexpress LLC Supplier No. HYN0473S850MG
L-Tyrosine-d2 is the deuterium labeled L-Tyrosine. L-Tyrosine is a non-essential amino acid which can inhibit citrate synthase activity in the posterior cortex. This product is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.