Learn More
Medchemexpress LLC L-tyrosine-d2 | 72963-27-0 | MFCD00144690 | 99.8% | 183.20 | C9H9D2NO3 | 10 MG
SDP
Catalog No. 5000295892
Encompass_Preferred
L-Tyrosine-d2 is the deuterium-labeled form of the amino acid L-tyrosine supplied as a white to off-white solid for analytical and research use. It is intended primarily as a tracer and as an internal standard for quantitative analyses and metabolic studies that require isotopically labeled material. The material is supplied in a 10 mg quantity and has a reported molecular formula C9H9D2NO3 and molecular weight 183.20.
  • Deuterium labeled (d2) isotopologue for tracer and standard use.
  • Suitable as an internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High reported purity (~99.8%) for analytical applications.
  • Solid appearance: white to off-white.
  • Recommended sealed storage away from moisture and light; in solvent: -80°C up to 6 months, -20°C up to 1 month.
Catalog No. 50-002-95892 Supplier Medchemexpress LLC Supplier No. HYN0473S810MG
L-Tyrosine-d2 is the deuterium-labeled form of the amino acid L-tyrosine supplied as a white to off-white solid for analytical and research use. It is intended primarily as a tracer and as an internal standard for quantitative analyses and metabolic studies that require isotopically labeled material. The material is supplied in a 10 mg quantity and has a reported molecular formula C9H9D2NO3 and molecular weight 183.20.
  • Deuterium labeled (d2) isotopologue for tracer and standard use.
  • Suitable as an internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High reported purity (~99.8%) for analytical applications.
  • Solid appearance: white to off-white.
  • Recommended sealed storage away from moisture and light; in solvent: -80°C up to 6 months, -20°C up to 1 month.