Learn More
Medchemexpress LLC L-Tyrosine-d3 | 71939-39-4 | 184.21 | 1 MG
SDP
Catalog No. 5000389782
Encompass_Preferred
L-Tyrosine-d3 is a deuterium-labeled form of L-Tyrosine, a non-essential amino acid. It is designed for research applications, including use as a tracer and internal standard.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-89782 Supplier Medchemexpress LLC Supplier No. HYN0473S151MG
L-Tyrosine-d3 is a deuterium-labeled form of L-Tyrosine, a non-essential amino acid. It is designed for research applications, including use as a tracer and internal standard.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules
  • Potential to affect pharmacokinetic and metabolic profiles of drugs