Learn More
Medchemexpress LLC L-Tyrosine-d3 | 71939-39-4 | 184.21 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0473S155MG
Encompass_Preferred
L-Tyrosine-d3 is a deuterium-labeled form of L-Tyrosine, a non-essential amino acid. It is utilized as a tracer and internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules primarily as tracers for quantitation during the drug development process.
  • Inhibits citrate synthase activity in the posterior cortex.
  • Used as a tracer for quantitation in drug development.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-89370