Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-tyrosine-d3 | 71939-39-4 | 99.0% | 184.21 g/mol | C9H8D3NO3 | 10 MG
SDP

Catalog No. 5000296378
Encompass_Preferred

L-Tyrosine-d3 is the deuterium-labeled form of L-tyrosine supplied as a solid stable isotope standard for analytical and research use. It is used as a tracer and internal standard in quantitative methods such as NMR, GC-MS, and LC-MS, and may be used in metabolic studies involving citrate synthase activity in the posterior cortex.

  • Deuterium-labeled tracer and internal standard for NMR, GC-MS, and LC-MS.
  • High purity (≈99.0%) suitable for analytical applications.
  • Solid, white to off-white appearance for easy handling.
  • Storage: powder stable at -20°C (3 years) and 4°C (2 years); in solvent store at -80°C (6 months) or -20°C (1 month).
  • Molecular weight 184.21 g/mol; formula C9H8D3NO3.

Catalog No. 50-002-96378 Supplier Medchemexpress LLC Supplier No. HYN0473S1510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Tyrosine-d3 is the deuterium-labeled form of L-tyrosine supplied as a solid stable isotope standard for analytical and research use. It is used as a tracer and internal standard in quantitative methods such as NMR, GC-MS, and LC-MS, and may be used in metabolic studies involving citrate synthase activity in the posterior cortex.

  • Deuterium-labeled tracer and internal standard for NMR, GC-MS, and LC-MS.
  • High purity (≈99.0%) suitable for analytical applications.
  • Solid, white to off-white appearance for easy handling.
  • Storage: powder stable at -20°C (3 years) and 4°C (2 years); in solvent store at -80°C (6 months) or -20°C (1 month).
  • Molecular weight 184.21 g/mol; formula C9H8D3NO3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.