Learn More
Medchemexpress LLC L-Tyrosine-d4 | 62595-14-6 | 185.21 | 50 MG
SDP
Catalog No. 5000389905
Encompass_Preferred
L-Tyrosine-d4 is a deuterium labeled non-essential amino acid. It inhibits citrate synthase activity in the posterior cortex and increases succinate dehydrogenase in various tissues. This compound is suitable for various quantitative analyses and research applications.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Inhibits citrate synthase activity
  • Increases succinate dehydrogenase activity in specific brain regions and liver
Catalog No. 50-003-89905 Supplier Medchemexpress LLC Supplier No. HYN0473S50MG
L-Tyrosine-d4 is a deuterium labeled non-essential amino acid. It inhibits citrate synthase activity in the posterior cortex and increases succinate dehydrogenase in various tissues. This compound is suitable for various quantitative analyses and research applications.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Inhibits citrate synthase activity
  • Increases succinate dehydrogenase activity in specific brain regions and liver