Learn More
Medchemexpress LLC L-Tyrosine-d4 | 62595-14-6 | MFCD00079605 | 185.21 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0473S5MG
Encompass_Preferred
L-Tyrosine-d4 is a deuterium labeled L-Tyrosine. L-Tyrosine is a non-essential amino acid which can inhibit citrate synthase activity in the posterior cortex.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89990