Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-tyrosine-d5 | 1226919-57-8 | MFCD18782928 | 99.9% | 186.22 | C9H6D5NO3 | 10 MG
SDP

Catalog No. 5000296269
Encompass_Preferred

L-Tyrosine-d5 is a deuterium-labeled stable isotope of the amino acid L-tyrosine supplied as a white to off-white powder for research use. It is intended for use as an internal standard or tracer in mass spectrometry and metabolic studies, providing high isotopic fidelity for quantitative analyses.

  • High isotopic and chemical purity for analytical standards and quantitative assays.
  • Supplied as a solid powder with defined storage recommendations for stability.
  • Suitable as an internal standard in LC-MS/MS and metabolic tracing workflows.
  • Soluble in acidic aqueous solutions; limited solubility in DMSO without ultrasonic assistance.
  • Recommended storage: powder at -20°C for long-term stability.

Catalog No. 50-002-96269 Supplier Medchemexpress LLC Supplier No. HYN0473S1410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Tyrosine-d5 is a deuterium-labeled stable isotope of the amino acid L-tyrosine supplied as a white to off-white powder for research use. It is intended for use as an internal standard or tracer in mass spectrometry and metabolic studies, providing high isotopic fidelity for quantitative analyses.

  • High isotopic and chemical purity for analytical standards and quantitative assays.
  • Supplied as a solid powder with defined storage recommendations for stability.
  • Suitable as an internal standard in LC-MS/MS and metabolic tracing workflows.
  • Soluble in acidic aqueous solutions; limited solubility in DMSO without ultrasonic assistance.
  • Recommended storage: powder at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.