Learn More
Medchemexpress LLC L-Tyrosine, methyl ester | 1080-06-4 | 195.22 | 25 G
SDP
Catalog No. 5000448150
Encompass_Preferred
H-Tyr-OMe is an endogenous metabolite. It quenches the fluorescence of carbon dots by oxidizing the phenolic hydroxyl group to quinone under the catalysis of Tyrosinase. Its amino and thiol binding ability can respond to changes in the concentration of biothiols.
  • Endogenous metabolite
  • Quenches fluorescence of carbon dots
  • Responds to changes in biothiol concentration
  • For research use only
Catalog No. 50-004-48150 Supplier Medchemexpress LLC Supplier No. 25GHYW007671
H-Tyr-OMe is an endogenous metabolite. It quenches the fluorescence of carbon dots by oxidizing the phenolic hydroxyl group to quinone under the catalysis of Tyrosinase. Its amino and thiol binding ability can respond to changes in the concentration of biothiols.
  • Endogenous metabolite
  • Quenches fluorescence of carbon dots
  • Responds to changes in biothiol concentration
  • For research use only