Learn More
Cayman Chemical L-TyrosInsodIum salt hydra 25g
SDP
Catalog No. 502911883
Encompass_Preferred
A conditionally essential amino acid; a precursor in the biosynthesis of catecholamine neurotransmitters, melanins, and thyroid hormones; plasma, skeletal muscle, and erythrocyte levels are decreased in patients with chronic kidney disease
Catalog No. 50-291-1883 Supplier Cayman Chemical Supplier No. 3633325 G
A conditionally essential amino acid; a precursor in the biosynthesis of catecholamine neurotransmitters, melanins, and thyroid hormones; plasma, skeletal muscle, and erythrocyte levels are decreased in patients with chronic kidney disease