Learn More
Medchemexpress LLC L-Uridine | 26287-69-4 | 244.20 | 250 MG
SDP
Catalog No. 5000449974
Encompass_Preferred
L-Uridine is an enantiomer of the normal RNA constituent D-uridine. It can be isolated from the Polyporaceae fungus *Poria cocos* (Schw.) and acts as a phosphate acceptor for nucleoside phosphotransferases. In vitro, L-uridine (7 min) is converted to 5'-L-uridylic acid, with a Km value of 28 mM.
  • Enantiomer of D-uridine
  • Isolated from *Poria cocos* (Schw.)
  • Acts as a phosphate acceptor for nucleoside phosphotransferases
  • High purity (99.81%)
  • Available as a solid
  • Soluble in various in vitro dissolution methods
  • Stable under specified storage conditions
Catalog No. 50-004-49974 Supplier Medchemexpress LLC Supplier No. 250MGHYW006429
L-Uridine is an enantiomer of the normal RNA constituent D-uridine. It can be isolated from the Polyporaceae fungus *Poria cocos* (Schw.) and acts as a phosphate acceptor for nucleoside phosphotransferases. In vitro, L-uridine (7 min) is converted to 5'-L-uridylic acid, with a Km value of 28 mM.
  • Enantiomer of D-uridine
  • Isolated from *Poria cocos* (Schw.)
  • Acts as a phosphate acceptor for nucleoside phosphotransferases
  • High purity (99.81%)
  • Available as a solid
  • Soluble in various in vitro dissolution methods
  • Stable under specified storage conditions