Learn More
Medchemexpress LLC L-valine-1,2,3,4,4′-13C5 | 55443-52-2 | MFCD00144707 | 99.6% | 122.11 g/mol | 13C5H11NO2 | 10 MG
SDP
Catalog No. 5000295952
Encompass_Preferred
L-Valine-13C5 is the 13C-labeled isotopologue of the amino acid L-valine, supplied as a white to off-white powder for research and analytical use. It is commonly used as an internal standard, tracer, or labeled substrate in metabolic and mass-spectrometry studies.
  • Isotopically labeled (13C5) for tracer and mass-spectrometry applications.
  • High purity (99.6%) appropriate for analytical work.
  • Solid powder form, convenient for weighing and dissolution.
  • Small pack size (10 mg) suitable for low-consumption experiments.
  • Storage recommendations: powder -20°C for long-term stability.
Catalog No. 50-002-95952 Supplier Medchemexpress LLC Supplier No. HYN0717S610MG
L-Valine-13C5 is the 13C-labeled isotopologue of the amino acid L-valine, supplied as a white to off-white powder for research and analytical use. It is commonly used as an internal standard, tracer, or labeled substrate in metabolic and mass-spectrometry studies.
  • Isotopically labeled (13C5) for tracer and mass-spectrometry applications.
  • High purity (99.6%) appropriate for analytical work.
  • Solid powder form, convenient for weighing and dissolution.
  • Small pack size (10 mg) suitable for low-consumption experiments.
  • Storage recommendations: powder -20°C for long-term stability.