Learn More
Medchemexpress LLC L-Valine-13C5,15N | 202407-30-5 | 123.10 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN0717S11MG
Encompass_Preferred
L-Valine-13C5,15N is the 13C- and 15N-labeled L-Valine. This nonlinear semiorganic material causes lipid peroxidation and accumulation of malondialdehyde (MDA), and exhibits inhibitory activity against cyanobacteria. It also inhibits multidrug-resistant bacteria through activation of the PI3K/Akt signaling pathway and inhibition of arginase.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements for tracing during drug development.
  • Exhibits inhibitory activity against cyanobacteria.
  • Inhibits multidrug-resistant bacteria.
Catalog No. 50-004-16749