Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Valine-13C5,15N | 202407-30-5 | 123.10 | 25 MG
SDP

Catalog No. 5000390323
Encompass_Preferred

L-Valine-13C5,15N is the 13C- and 15N-labeled L-Valine. It is a nonlinear semiorganic material that causes lipid peroxidation and accumulation of malondialdehyde (MDA), and exhibits inhibitory activity against cyanobacteria. This compound inhibits multidrug-resistant bacteria through activation of PI3K/Akt signaling pathway and inhibition of arginase. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.

  • Purity: 99.64%
  • Appearance: Solid
  • Color: White to off-white
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS

Catalog No. 50-003-90323 Supplier Medchemexpress LLC Supplier No. 25MGHYN0717S1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Valine-13C5,15N is the 13C- and 15N-labeled L-Valine. It is a nonlinear semiorganic material that causes lipid peroxidation and accumulation of malondialdehyde (MDA), and exhibits inhibitory activity against cyanobacteria. This compound inhibits multidrug-resistant bacteria through activation of PI3K/Akt signaling pathway and inhibition of arginase. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.

  • Purity: 99.64%
  • Appearance: Solid
  • Color: White to off-white
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.