Learn More
Medchemexpress LLC L-Valine-13C5,15N | 202407-30-5 | 123.10 | 5 MG
SDP
Catalog No. 5000416637
Encompass_Preferred
L-Valine-13C5,15N is a 13C- and 15N-labeled L-Valine, a nonlinear semiorganic material. It causes lipid peroxidation, accumulates malondialdehyde (MDA), and inhibits cyanobacteria. It also inhibits multidrug-resistant bacteria by activating the PI3K/Akt pathway and inhibiting arginase. This compound serves as a tracer and an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS, utilizing stable heavy isotopes for drug development and pharmacokinetic studies.
  • 13C- and 15N-labeled L-Valine
  • Inhibits cyanobacteria and multidrug-resistant bacteria
  • Functions as a tracer
  • Acts as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Utilizes stable heavy isotopes for drug development and pharmacokinetic studies
Catalog No. 50-004-16637 Supplier Medchemexpress LLC Supplier No. HYN0717S15MG
L-Valine-13C5,15N is a 13C- and 15N-labeled L-Valine, a nonlinear semiorganic material. It causes lipid peroxidation, accumulates malondialdehyde (MDA), and inhibits cyanobacteria. It also inhibits multidrug-resistant bacteria by activating the PI3K/Akt pathway and inhibiting arginase. This compound serves as a tracer and an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS, utilizing stable heavy isotopes for drug development and pharmacokinetic studies.
  • 13C- and 15N-labeled L-Valine
  • Inhibits cyanobacteria and multidrug-resistant bacteria
  • Functions as a tracer
  • Acts as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Utilizes stable heavy isotopes for drug development and pharmacokinetic studies