Learn More
Medchemexpress LLC L-valine (13C5,15N) | 202407-30-5 | MFCD00144708 | 98.0% | 123.10 g·mol⁻1 | 13C5H11 15N O2 | 10 MG
SDP
Catalog No. 5000296607
Encompass_Preferred
L-valine labeled with five 13C atoms and one 15N atom, supplied as a white to off-white powder for analytical and tracer applications. It is intended for use in mass spectrometry, metabolic labeling, SILAC, and other experiments requiring isotopically labeled amino acids. Store the powder at low temperature and follow solvent storage guidelines for solutions.
Catalog No. 50-002-96607 Supplier Medchemexpress LLC Supplier No. HYN0717S110MG
L-valine labeled with five 13C atoms and one 15N atom, supplied as a white to off-white powder for analytical and tracer applications. It is intended for use in mass spectrometry, metabolic labeling, SILAC, and other experiments requiring isotopically labeled amino acids. Store the powder at low temperature and follow solvent storage guidelines for solutions.