Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals L-Valine, 99%

Catalog No. AC140811000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC140811000 100 g Plastic Bottle
AC140810250 25 g Plastic Bottle
AC140815000 500 g Plastic Bottle
AC140810025 2.5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC140811000 Supplier Thermo Scientific Chemicals Supplier No. 140811000
  • $130.70
    $95.65 / Each of 1

    Save $35.05

    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

CAS: 72-18-4 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-18-4
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064220
InChI Key KZSNJWFQEVHDMF-BYPYZUCNSA-N
Synonym l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin
PubChem CID 6287
ChEBI CHEBI:16414
IUPAC Name (2S)-2-amino-3-methylbutanoic acid
SMILES CC(C)[C@H](N)C(O)=O

Specifications

Melting Point 315.0°C
Ammonium 200ppm max.
Color White
Chlorine (Cl) 200ppm max.
Foreign Amino Acids 0.5% max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Loss on Drying 0.2% max.
Assay Percent Range 99%
Packaging Plastic Bottle
Linear Formula (CH3)2CHCH(NH2)CO2H
Specific Rotation + 28.00
Beilstein 04, 427
Fieser 13,226
Sulfate 300ppm max.
Merck Index 15, 10095
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 85g/L (20°C). Other solubilities: practically insoluble in cold alcohol, ether and, acetone, soluble in mineral acids, insoluble in chloroform, diethylether
Density 1.2300g/mL
Specific Rotation Condition +28.00 (20.00°C c=8,6N HCl)
Quantity 100 g
Formula Weight 117.15
Percent Purity 99%
Specific Gravity 1.23
Physical Form Powder
Chemical Name or Material L-Valine, 99%
Show More Show Less
EINECSNumber 200-773-6
RTECSNumber YV9361000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.