Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Valine benzyl ester hydrochloride, 95%

Catalog No. AAH6224206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6224206 5 g
AAH6224214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6224206 Supplier Thermo Scientific Chemicals Supplier No. H6224206
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2462-34-2 | C12H18ClNO2 | 243.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2462-34-2
Molecular Formula C12H18ClNO2
Molecular Weight (g/mol) 243.73
MDL Number MFCD22422648
InChI Key ZIUNABFUHGBCMF-MERQFXBCSA-N
Synonym l-valine benzyl ester hydrochloride, val-obzl hcl, h-val-obzl.hcl, s-benzyl 2-amino-3-methylbutanoate hydrochloride, valine benzyl ester hydrochloride, benzyl 2s-2-amino-3-methylbutanoate hydrochloride, val-obzl.hcl, val-obzl. hcl, l-valine benzyl ester hydrochloride
PubChem CID 11601206
IUPAC Name benzyl (2S)-2-amino-3-methylbutanoate;hydrochloride
SMILES Cl.CC(C)[C@H](N)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 138°C
Quantity 5 g
Formula Weight 243.73
Optical Rotation −10° (c=2 in Water)
Percent Purity 95%
Chemical Name or Material L-Valine benzyl ester hydrochloride
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.