Learn More
Medchemexpress LLC L-Valine-d1 | 77257-03-5 | 118.15 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0717S8
Encompass_Preferred
L-Valine-d1 is a deuterium-labeled version of L-Valine. L-Valine is a nonlinear semiorganic material that can cause lipid peroxidation and malondialdehyde (MDA) accumulation, and it has inhibitory activity against cyanobacteria. It also inhibits multidrug-resistant bacteria by activating the PI3K/Akt signaling pathway and inhibiting arginase.
  • Can be used as a tracer and as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Stable heavy isotope where hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-89277