Learn More
Medchemexpress LLC L-valine-d1 | 77257-03-5 | 118.15 g/mol | C5H10DNO2 | 10 MG
SDP
Catalog No. 5000296187
Encompass_Preferred
L-Valine-d1 is the deuterium-labeled analogue of L-valine supplied as a solid research reagent. It is intended for laboratory and analytical use, including mass spectrometry internal standards and metabolic tracing.
  • Single deuterium substitution provides a predictable mass shift for MS-based quantitation.
  • Suitable as an internal standard and tracer in metabolic and analytical studies.
  • Supplied as a solid powder (10 mg) for convenient storage and handling.
  • Stable when stored as powder at -20°C; in solvent, store at -80°C for long-term stability.
  • Intended for research use only.
Catalog No. 50-002-96187 Supplier Medchemexpress LLC Supplier No. HYN0717S810MG
L-Valine-d1 is the deuterium-labeled analogue of L-valine supplied as a solid research reagent. It is intended for laboratory and analytical use, including mass spectrometry internal standards and metabolic tracing.
  • Single deuterium substitution provides a predictable mass shift for MS-based quantitation.
  • Suitable as an internal standard and tracer in metabolic and analytical studies.
  • Supplied as a solid powder (10 mg) for convenient storage and handling.
  • Stable when stored as powder at -20°C; in solvent, store at -80°C for long-term stability.
  • Intended for research use only.