Learn More
Medchemexpress LLC L-Valine-d1 ((S)-Valine-d1) | 77257-03-5 | 118.15 | 100 MG
SDP
Catalog No. 5000416703
Encompass_Preferred
L-Valine-d1 is the deuterium-labeled version of L-Valine. L-Valine itself is a nonlinear semiorganic material known to induce lipid peroxidation and malondialdehyde accumulation, and to inhibit cyanobacteria. It also inhibits multidrug-resistant bacteria by activating the PI3K/Akt signaling pathway and suppressing arginase.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis via techniques such as NMR, GC-MS, or LC-MS.
Catalog No. 50-004-16703 Supplier Medchemexpress LLC Supplier No. HYN0717S8100MG
L-Valine-d1 is the deuterium-labeled version of L-Valine. L-Valine itself is a nonlinear semiorganic material known to induce lipid peroxidation and malondialdehyde accumulation, and to inhibit cyanobacteria. It also inhibits multidrug-resistant bacteria by activating the PI3K/Akt signaling pathway and suppressing arginase.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis via techniques such as NMR, GC-MS, or LC-MS.