Learn More
Medchemexpress LLC L-valine-d8 | 35045-72-8 | MFCD00144704 | 99.8% | 125.20 g·mol⁻¹ | C5H3D8NO2 | 10 MG
SDP
Catalog No. 5000297210
Encompass_Preferred
L-Valine-d8 is the deuterated isotopologue of the essential amino acid L-valine, labeled with eight deuterium atoms and supplied for research use as a stable isotope standard or for labeled synthesis. It is commonly used as an internal standard in mass spectrometry and for metabolic and isotopic-labeling studies.
  • Deuterated with eight deuterium atoms for mass-difference applications.
  • High isotopic enrichment, suitable for quantitative MS analysis.
  • High chemical purity for research applications.
  • Available in small solid vials and solution formats for flexible use.
Catalog No. 50-002-97210 Supplier Medchemexpress LLC Supplier No. HYI112410MG
L-Valine-d8 is the deuterated isotopologue of the essential amino acid L-valine, labeled with eight deuterium atoms and supplied for research use as a stable isotope standard or for labeled synthesis. It is commonly used as an internal standard in mass spectrometry and for metabolic and isotopic-labeling studies.
  • Deuterated with eight deuterium atoms for mass-difference applications.
  • High isotopic enrichment, suitable for quantitative MS analysis.
  • High chemical purity for research applications.
  • Available in small solid vials and solution formats for flexible use.