Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-valine, N-[4-(4-propylcyclohexyl)benzoyl]- | 1241902-40-8 | 99.8% | 345.48 g/mol | C21H31NO3 | 100MG
SDP

Catalog No. 5000236616
Encompass_Preferred

GNE-0439 is a small-molecule Nav1.7-selective sodium channel inhibitor used in research to study voltage-gated sodium channel pharmacology. It contains a carboxylic acid moiety, exhibits Nav1.7 IC50 = 0.34 μM with reduced activity at Nav1.5 (IC50 = 38.3 μM), and has molecular formula C21H31NO3 (MW 345.48 g/mol).

  • Nav1.7-selective sodium channel inhibitor with Nav1.7 IC50 0.34 μM.
  • Shows reduced activity on Nav1.5 (IC50 38.3 μM), indicating selectivity.
  • Molecular formula C21H31NO3 and molecular weight 345.48 g/mol.
  • High purity (99.8%) suitable for research applications.
  • Soluble in DMSO (~130 mg/mL) and compatible with common in vivo vehicles.
  • Solid is stable at 4°C; solutions are recommended at -80°C for long-term storage.
  • Offered in solid and solution formats for flexible dosing and formulation.

Catalog No. 50-002-36616 Supplier Medchemexpress LLC Supplier No. HY123824100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GNE-0439 is a small-molecule Nav1.7-selective sodium channel inhibitor used in research to study voltage-gated sodium channel pharmacology. It contains a carboxylic acid moiety, exhibits Nav1.7 IC50 = 0.34 μM with reduced activity at Nav1.5 (IC50 = 38.3 μM), and has molecular formula C21H31NO3 (MW 345.48 g/mol).

  • Nav1.7-selective sodium channel inhibitor with Nav1.7 IC50 0.34 μM.
  • Shows reduced activity on Nav1.5 (IC50 38.3 μM), indicating selectivity.
  • Molecular formula C21H31NO3 and molecular weight 345.48 g/mol.
  • High purity (99.8%) suitable for research applications.
  • Soluble in DMSO (~130 mg/mL) and compatible with common in vivo vehicles.
  • Solid is stable at 4°C; solutions are recommended at -80°C for long-term storage.
  • Offered in solid and solution formats for flexible dosing and formulation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.