Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-valine, N,N-dimethyl- | 2812-32-0 | ≥98.0% | 25 G
SDP

Catalog No. 5000233266
Encompass_Preferred

N,N-dimethyl-L-valine is an N,N-dimethylated derivative of the amino acid L-valine used as a synthetic intermediate in pharmaceutical and medicinal chemistry. It is supplied as a white to off-white solid, typically ≥98.0% pure.

  • Used as a synthetic intermediate in pharmaceutical synthesis.
  • Non-proteinogenic amino acid derivative.
  • Purity ≥98.0% and molecular weight 145.20 g·mol⁻¹.
  • Physical state: white to off-white solid.
  • Available in laboratory pack sizes, including 25 G.

Catalog No. 50-002-33266 Supplier Medchemexpress LLC Supplier No. HY11941425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N,N-dimethyl-L-valine is an N,N-dimethylated derivative of the amino acid L-valine used as a synthetic intermediate in pharmaceutical and medicinal chemistry. It is supplied as a white to off-white solid, typically ≥98.0% pure.

  • Used as a synthetic intermediate in pharmaceutical synthesis.
  • Non-proteinogenic amino acid derivative.
  • Purity ≥98.0% and molecular weight 145.20 g·mol⁻¹.
  • Physical state: white to off-white solid.
  • Available in laboratory pack sizes, including 25 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.