Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-valine, N,N-dimethyl- | 2812-32-0 | ≥98.0% | 50 G
SDP

Catalog No. 5000234275
Encompass_Preferred

N,N-dimethyl-L-valine is a synthetic, non-proteinogenic amino acid derivative used as a building block and intermediate in pharmaceutical and organic synthesis. It is supplied as a white to off-white solid with high purity suitable for research and synthetic applications.

  • CAS number 2812-32-0
  • Molecular formula C7H15NO2
  • Molecular weight 145.20 g/mol
  • Purity ≥98.0%
  • White to off-white solid appearance
  • Typical use as a synthetic intermediate in drug and organic synthesis

Catalog No. 50-002-34275 Supplier Medchemexpress LLC Supplier No. HY11941450G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N,N-dimethyl-L-valine is a synthetic, non-proteinogenic amino acid derivative used as a building block and intermediate in pharmaceutical and organic synthesis. It is supplied as a white to off-white solid with high purity suitable for research and synthetic applications.

  • CAS number 2812-32-0
  • Molecular formula C7H15NO2
  • Molecular weight 145.20 g/mol
  • Purity ≥98.0%
  • White to off-white solid appearance
  • Typical use as a synthetic intermediate in drug and organic synthesis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.