Learn More
Alkali Scientific L Valine, USP Grade, 1Kg
SDP
Catalog No. 5000661111
Encompass_Preferred
L Valine, USP Grade, available in a 1kg package, is a high-purity amino acid essential for protein synthesis and muscle metabolism. It plays a key role in the synthesis of proteins and helps regulate blood sugar levels. Valine is also important for maintaining nitrogen balance in the body, making it an essential amino acid in clinical nutrition and medical formulations. The USP grade ensures the highest standards of purity, making it suitable for use in laboratory assays, clinical studies, and pharmaceutical applications. This amino acid is also used in the formulation of dietary supplements and in metabolic research.
Catalog No. 50-006-61111 Supplier Alkali Scientific Supplier No. LVA002CP
L Valine, USP Grade, available in a 1kg package, is a high-purity amino acid essential for protein synthesis and muscle metabolism. It plays a key role in the synthesis of proteins and helps regulate blood sugar levels. Valine is also important for maintaining nitrogen balance in the body, making it an essential amino acid in clinical nutrition and medical formulations. The USP grade ensures the highest standards of purity, making it suitable for use in laboratory assays, clinical studies, and pharmaceutical applications. This amino acid is also used in the formulation of dietary supplements and in metabolic research.