Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Valine, USP, JP, bioCERTIFIED, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18614865 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 kg
12 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-865 12 kg Plastic Bottle
18-614-864 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-614-865 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. V910012KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Spectrum™ Chemical bioCERTIFIED products are suitable for biopharmaceutical production.

  • Full testing including bioburden, endotoxin and elemental analysis
  • Comprehensive regulatory and scientific documentation
  • Change control and batch traceabilitiy
  • Biopharm industry standard CFR 210/211 cGMP

Chemical Identifiers

CAS 72-18-4

Specifications

Ammonium 0.02%
Arsenic (As) 2 ppm
Bacterial Endotoxins 6.0 EU/g
Chlorine (Cl) 0.021%
Heavy Metals (as Pb) 20 ppm
Identification To pass test
Iron (Fe) 10 ppm
Loss on Drying 0.2%
Packaging Plastic Bottle
Residual Solvents To pass test
Residue after Ignition 0.1%
Sulfate 0.03%
Molecular Weight (g/mol) 117.15
Quantity 12 kg
Percent Purity 98.5 to 101%
Grade USP/JP/bioCERTIFIED
Chemical Name or Material L-Valine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.