Learn More
Cayman Chemical L-Valine-d8, Purity: ≥99% deuterated forms (d1-d8), CAS: 35045-72-8, 10 mg
SDP
Catalog No. 502913114
Encompass_Preferred
An internal standard for the quantification of L-valine by GC- or LC-MS
Catalog No. 50-291-3114 Supplier Cayman Chemical Supplier No. 3483010 MG
An internal standard for the quantification of L-valine by GC- or LC-MS